siRNA Details
| Gene Symbol: Brix1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029915.1 | NP_001025086.1 | 10 | 1803 | ||
| GenBank mRNA | BC098005.1 | - | 10 | 1803 | 7453609 | |
| FQ213839.1 | - | 1 | 331 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Brix1| Gene Symbol: Brix1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029915.1 | NP_001025086.1 | 10 | 1803 | ||
| GenBank mRNA | BC098005.1 | - | 10 | 1803 | 7453609 | |
| FQ213839.1 | - | 1 | 331 | |||