siRNA Details
| Gene Symbol: Mrpl47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037183.1 | NP_001032260.1 | 7 | 1583 | ||
| GenBank mRNA | BC089884.1 | - | 7 | 1584 | ||
| BC105820.1 | - | 7 | 1583 | 7369885 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mrpl47| Gene Symbol: Mrpl47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037183.1 | NP_001032260.1 | 7 | 1583 | ||
| GenBank mRNA | BC089884.1 | - | 7 | 1584 | ||
| BC105820.1 | - | 7 | 1583 | 7369885 | ||