siRNA Details
| Gene Symbol: RGD1307100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037184.1 | NP_001032261.1 | 3, 4 | 792 | ||
| XM_006232275.2 | XP_006232337.1 | 87, 88 | 15252 | |||
| XM_008760914.1 | XP_008759136.1 | 87, 88 | 15252 | |||
| XM_008760915.1 | XP_008759137.1 | 87, 88 | 15249 | |||
| XM_008760916.1 | XP_008759138.1 | 86, 87 | 15201 | |||
| XM_008760917.1 | XP_008759139.1 | 86, 87 | 15189 | |||
| XM_008760918.1 | XP_008759140.1 | 86, 87 | 15114 | |||
| XM_008760919.1 | XP_008759141.1 | 86, 87 | 15050 | |||
| GenBank mRNA | BC100102.1 | - | 3, 4 | 792 | 7377390 | |

RGD1307100
Genomic Map