siRNA Details
| Gene Symbol: Pecam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031591.1 | NP_113779.1 | 280 | |||
| GenBank mRNA | BC104711.1 | - | 280 | 7368518 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pecam1| Gene Symbol: Pecam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031591.1 | NP_113779.1 | 280 | |||
| GenBank mRNA | BC104711.1 | - | 280 | 7368518 | ||