siRNA Details
| Gene Symbol: Depdc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029916.1 | NP_001025087.1 | 4, 5 | 834 | ||
| XM_006234642.2 | XP_006234704.1 | 4, 5 | 857 | |||
| GenBank mRNA | BC097357.1 | - | 4, 5 | 834 | 7442188 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Depdc7| Gene Symbol: Depdc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029916.1 | NP_001025087.1 | 4, 5 | 834 | ||
| XM_006234642.2 | XP_006234704.1 | 4, 5 | 857 | |||
| GenBank mRNA | BC097357.1 | - | 4, 5 | 834 | 7442188 | |