siRNA Details
| Gene Symbol: Jph2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037974.1 | NP_001033063.1 | 1 | 344 | ||
| XM_006235526.2 | XP_006235588.1 | 1 | 1235 | |||
| GenBank mRNA | DQ304564.1 | - | 1 | 344 | ||
| FQ223967.1 | - | 1 | 771 | |||

Jph2
Genomic Map