siRNA Details
| Gene Symbol: Slc25a40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037186.1 | NP_001032263.1 | 781 | |||
| GenBank mRNA | BC100071.1 | - | 781 | 7322585 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc25a40| Gene Symbol: Slc25a40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037186.1 | NP_001032263.1 | 781 | |||
| GenBank mRNA | BC100071.1 | - | 781 | 7322585 | ||