siRNA Details
Gene Symbol: Iqub | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034130.1 | NP_001029302.1 | 10 | 1652 | ||
GenBank mRNA | BC099086.1 | - | 10 | 1652 | 7454794 | |
DQ132434.1 | - | 10 | 1699 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Iqub | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034130.1 | NP_001029302.1 | 10 | 1652 | ||
GenBank mRNA | BC099086.1 | - | 10 | 1652 | 7454794 | |
DQ132434.1 | - | 10 | 1699 |