siRNA Details
| Gene Symbol: Mob1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033891.1 | NP_001029063.1 | 5 | 723 | ||
| GenBank mRNA | BC101879.1 | - | 5 | 723 | 7929996 | |
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_145571, BC033463, AK167515, AK172205, AK158228, and others.

Mob1a
Genomic Map