siRNA Details
Gene Symbol: Nagk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037768.1 | NP_001032857.1 | 10 | 1175 | ||
GenBank mRNA | BC107647.1 | - | 10 | 1175 | 7930414 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nagk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037768.1 | NP_001032857.1 | 10 | 1175 | ||
GenBank mRNA | BC107647.1 | - | 10 | 1175 | 7930414 |