siRNA Details
| Gene Symbol: Slc31a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC099751.1 | - | 3 | 793 | 5623519 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc31a2| Gene Symbol: Slc31a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC099751.1 | - | 3 | 793 | 5623519 | |