siRNA Details
| Gene Symbol: RGD1559786 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034132.1 | NP_001029304.1 | 8 | 757 | ||
| XM_006238508.2 | XP_006238570.1 | 8 | 873 | |||
| XM_006238509.2 | XP_006238571.1 | 7 | 948 | |||
| XM_008763867.1 | XP_008762089.1 | 7 | 716 | |||
| GenBank mRNA | BC100100.1 | - | 8 | 757 | 7376782 | |
| FQ228722.1 | - | 8 | 776 | |||

RGD1559786
Genomic Map