siRNA Details
| Gene Symbol: Zfp36l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001036626.1 | NP_001031703.1 | 2 | 1989 | ||
| XM_008764455.1 | XP_008762677.1 | 2 | 4053 | |||
| Gene Symbol: LOC100911319 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750261.1 | XP_003750309.1 | 2 | 1989 | ||
| XM_008765049.1 | XP_008763271.1 | 2 | 2018 | |||

Zfp36l2
Genomic Map