siRNA Details
| Gene Symbol: Pqlc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034952.1 | NP_001030124.1 | 7 | 827 | ||
| GenBank mRNA | BC081879.1 | - | 5 | 1982 | 7108131 | |
| CK477351.1 | - | 7 | 650 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pqlc3| Gene Symbol: Pqlc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034952.1 | NP_001030124.1 | 7 | 827 | ||
| GenBank mRNA | BC081879.1 | - | 5 | 1982 | 7108131 | |
| CK477351.1 | - | 7 | 650 | |||