siRNA Details
| Gene Symbol: Golga5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033065.1 | NP_001028237.1 | 1 | 37 | ||
| XM_006240461.2 | XP_006240523.1 | 2 | 289 | |||
| GenBank mRNA | AY144587.1 | - | 1 | 37 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Golga5| Gene Symbol: Golga5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033065.1 | NP_001028237.1 | 1 | 37 | ||
| XM_006240461.2 | XP_006240523.1 | 2 | 289 | |||
| GenBank mRNA | AY144587.1 | - | 1 | 37 | ||