siRNA Details
| Gene Symbol: L3mbtl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033695.1 | NP_001028867.1 | 17 | 3260 | ||
| XM_008765711.1 | XP_008763933.1 | 17 | 3288 | |||
| GenBank mRNA | BC088331.1 | - | 4 | 1664 | ||
| BC101865.1 | - | 17 | 3260 | 7938856 | ||

L3mbtl2
Genomic Map