siRNA Details
| Gene Symbol: Fbxo22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037770.1 | NP_001032859.1 | 2, 3 | 301 | ||
| GenBank mRNA | BC087061.1 | - | 2, 3 | 331 | ||
| BC107670.1 | - | 2, 3 | 301 | 7937607 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fbxo22| Gene Symbol: Fbxo22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037770.1 | NP_001032859.1 | 2, 3 | 301 | ||
| GenBank mRNA | BC087061.1 | - | 2, 3 | 331 | ||
| BC107670.1 | - | 2, 3 | 301 | 7937607 | ||