siRNA Details
| Gene Symbol: Cep63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037772.1 | NP_001032861.2 | 11 | 1403 | ||
| NM_001244805.1 | NP_001231734.1 | 11 | 1572 | |||
| XM_008766490.1 | XP_008764712.1 | 13 | 1694 | |||
| XM_008766491.1 | XP_008764713.1 | 12 | 1418 | |||
| XM_008766492.1 | XP_008764714.1 | 11 | 2993 | |||
| XM_008766493.1 | XP_008764715.1 | 13 | 1569 | |||
| GenBank mRNA | BC083632.1 | - | 14 | 1656 | ||
| BC098948.1 | - | 11 | 1403 | |||
| BC161815.1 | - | 11 | 1572 | 9031816 | ||

Cep63
Genomic Map