siRNA Details
| Gene Symbol: Sp110 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034137.1 | NP_001029309.1 | 10 | 1175 | ||
| XM_006245370.2 | XP_006245432.1 | 9 | 1056 | |||
| XM_008767234.1 | XP_008765456.1 | 11 | 1239 | |||
| GenBank mRNA | BC105751.1 | - | 10 | 1175 | 5599224 | |

Sp110
Genomic Map