siRNA Details
| Gene Symbol: C1galt1c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001030033.1 | NP_001025204.1 | 2 | 344 | ||
| XM_006257487.1 | XP_006257549.1 | 2 | 393 | |||
| GenBank mRNA | BC099818.1 | - | 2 | 344 | 7930810 | |
| FQ209423.1 | - | 2 | 370 | |||

C1galt1c1
Genomic Map