siRNA Details
| Gene Symbol: Txlng | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037187.1 | NP_001032264.1 | 5 | 1254 | ||
| GenBank mRNA | BC099110.1 | - | 5 | 1254 | 7461546 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Txlng| Gene Symbol: Txlng | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037187.1 | NP_001032264.1 | 5 | 1254 | ||
| GenBank mRNA | BC099110.1 | - | 5 | 1254 | 7461546 | |