siRNA Details
| Gene Symbol: Mbtps2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001035007.1 | NP_001030179.1 | 11 | 1775 | ||
| XM_006256944.2 | XP_006257006.1 | 9 | 1464 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mbtps2| Gene Symbol: Mbtps2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001035007.1 | NP_001030179.1 | 11 | 1775 | ||
| XM_006256944.2 | XP_006257006.1 | 9 | 1464 | |||