siRNA Details
| Gene Symbol: Spdl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034138.1 | NP_001029310.1 | 2, 3 | 298 | ||
| XM_006246104.1 | XP_006246166.1 | 2, 3 | 336 | |||
| GenBank mRNA | BC105812.1 | - | 2, 3 | 298 | 7112484 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spdl1| Gene Symbol: Spdl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034138.1 | NP_001029310.1 | 2, 3 | 298 | ||
| XM_006246104.1 | XP_006246166.1 | 2, 3 | 336 | |||
| GenBank mRNA | BC105812.1 | - | 2, 3 | 298 | 7112484 | |