siRNA Details
| Gene Symbol: LOC303448 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037190.1 | NP_001032267.1 | 2 | 2280 | ||
| GenBank mRNA | BC100150.1 | - | 2 | 2280 | 7455257 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC303448| Gene Symbol: LOC303448 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037190.1 | NP_001032267.1 | 2 | 2280 | ||
| GenBank mRNA | BC100150.1 | - | 2 | 2280 | 7455257 | |