siRNA Details
| Gene Symbol: St6galnac2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031652.1 | NP_001026822.1 | 9 | 1192 | ||
| GenBank mRNA | BC099812.1 | - | 9 | 1192 | 7929942 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

St6galnac2| Gene Symbol: St6galnac2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031652.1 | NP_001026822.1 | 9 | 1192 | ||
| GenBank mRNA | BC099812.1 | - | 9 | 1192 | 7929942 | |