siRNA Details
| Gene Symbol: Ccdc130 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037644.1 | NP_001032733.1 | 9 | 1209 | ||
| XM_006255252.2 | XP_006255314.1 | 10 | 1320 | |||
| XM_006255253.2 | XP_006255315.1 | 10 | 1346 | |||
| GenBank mRNA | BC107912.1 | - | 9 | 1209 | 7460239 | |

Ccdc130
Genomic Map