siRNA Details
| Gene Symbol: Tmem177 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037776.1 | NP_001032865.1 | 2 | 778 | ||
| XM_006249664.2 | XP_006249726.1 | 2 | 1025 | |||
| XM_006249665.2 | XP_006249727.1 | 3 | 917 | |||
| GenBank mRNA | BC098688.1 | - | 2 | 778 | 7386020 | |

Tmem177
Genomic Map