siRNA Details
| Gene Symbol: Lrp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037777.1 | NP_001032866.1 | 5 | 1114 | ||
| GenBank mRNA | BC100651.1 | - | 5 | 1114 | 7932904 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrp10| Gene Symbol: Lrp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037777.1 | NP_001032866.1 | 5 | 1114 | ||
| GenBank mRNA | BC100651.1 | - | 5 | 1114 | 7932904 | |