siRNA Details
| Gene Symbol: Nkx3-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034144.1 | NP_001029316.1 | 2 | 509 | ||
| GenBank mRNA | BC100628.1 | - | 2 | 509 | 7938032 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nkx3-1| Gene Symbol: Nkx3-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034144.1 | NP_001029316.1 | 2 | 509 | ||
| GenBank mRNA | BC100628.1 | - | 2 | 509 | 7938032 | |