siRNA Details
| Gene Symbol: Ankrd16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033698.1 | NP_001028870.1 | 1336 | |||
| GenBank mRNA | BC099837.1 | - | 1336 | 7936269 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ankrd16| Gene Symbol: Ankrd16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033698.1 | NP_001028870.1 | 1336 | |||
| GenBank mRNA | BC099837.1 | - | 1336 | 7936269 | ||