siRNA Details
| Gene Symbol: Sh3rf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034187.1 | NP_001029359.1 | 4 | 1072 | ||
| GenBank mRNA | BC100155.1 | - | 4 | 1072 | 7461488 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sh3rf2| Gene Symbol: Sh3rf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034187.1 | NP_001029359.1 | 4 | 1072 | ||
| GenBank mRNA | BC100155.1 | - | 4 | 1072 | 7461488 | |