siRNA Details
| Gene Symbol: Elp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034145.1 | NP_001029317.1 | 21 | 2292 | ||
| GenBank mRNA | BC092606.1 | - | 10 | 1069 | ||
| BC100661.1 | - | 21 | 2292 | 7935317 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Elp2| Gene Symbol: Elp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034145.1 | NP_001029317.1 | 21 | 2292 | ||
| GenBank mRNA | BC092606.1 | - | 10 | 1069 | ||
| BC100661.1 | - | 21 | 2292 | 7935317 | ||