siRNA Details
| Gene Symbol: Zfp518a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001030038.1 | NP_001025209.1 | 5 | 1082 | ||
| XM_006231397.2 | XP_006231459.1 | 5 | 1260 | |||
| XM_006231398.2 | XP_006231460.1 | 5 | 1300 | |||
| XM_008760440.1 | XP_008758662.1 | 5 | 1396 | |||
| GenBank mRNA | BC099801.1 | - | 5 | 1082 | 7936437 | |

Zfp518a
Genomic Map