siRNA Details
| Gene Symbol: Atad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001035002.1 | NP_001030174.1 | 4 | 391 | ||
| XM_008760330.1 | XP_008758552.1 | 4 | 476 | |||
| GenBank mRNA | BC094514.1 | - | 4 | 391 | 7307334 | |
| EF688601.1 | - | 3 | 206 | |||

Atad1
Genomic Map