siRNA Details
| Gene Symbol: Atad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001035002.1 | NP_001030174.1 | 9 | 984 | ||
| XM_008760330.1 | XP_008758552.1 | 9 | 1069 | |||
| GenBank mRNA | AY325215.1 | - | 1537 | |||
| BC094514.1 | - | 9 | 984 | 7307334 | ||
| EF688601.1 | - | 8 | 799 | |||

Atad1
Genomic Map