siRNA Details
| Gene Symbol: Fam105a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037648.1 | NP_001032737.1 | 5 | 560 | ||
| GenBank mRNA | BC107649.1 | - | 5 | 560 | 7930470 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam105a| Gene Symbol: Fam105a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037648.1 | NP_001032737.1 | 5 | 560 | ||
| GenBank mRNA | BC107649.1 | - | 5 | 560 | 7930470 | |