siRNA Details
| Gene Symbol: Crtc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033895.1 | NP_001029067.1 | 10 | 931 | ||
| XM_008761146.1 | XP_008759368.1 | 9 | 976 | |||
| GenBank mRNA | BC128708.1 | - | 10 | 1056 | ||
| BC168242.1 | - | 10 | 1044 | 7934746 | ||
| DQ185515.1 | - | 10 | 931 | |||

Crtc2
Genomic Map