siRNA Details
| Gene Symbol: Crtc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033895.1 | NP_001029067.1 | 14 | 1878 | ||
| XM_008761146.1 | XP_008759368.1 | 13 | 1923 | |||
| GenBank mRNA | BC128708.1 | - | 14 | 2003 | ||
| BC168242.1 | - | 14 | 1991 | 7934746 | ||
| DQ185515.1 | - | 14 | 1878 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_028881, BC023831, AA276938, BC033300, AK079382, and others.

Crtc2
Genomic Map