siRNA Details
| Gene Symbol: Rassf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037096.1 | NP_001032173.1 | 11 | 2190 | ||
| XM_006235066.2 | XP_006235128.1 | 11 | 2873 | |||
| GenBank mRNA | BC107656.1 | - | 11 | 2190 | 7935449 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rassf2| Gene Symbol: Rassf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037096.1 | NP_001032173.1 | 11 | 2190 | ||
| XM_006235066.2 | XP_006235128.1 | 11 | 2873 | |||
| GenBank mRNA | BC107656.1 | - | 11 | 2190 | 7935449 | |