siRNA Details
| Gene Symbol: Slc52a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037198.1 | NP_001032275.1 | 4 | 1672 | ||
| XM_006235238.2 | XP_006235300.1 | 7 | 2343 | |||
| XM_008762327.1 | XP_008760549.1 | 7 | 2723 | |||
| XM_008762329.1 | XP_008760551.1 | 3 | 2047 | |||
| GenBank mRNA | BC099097.1 | - | 4 | 1672 | 7459579 | |
More Information
Set Membership: |
|

Slc52a3
Genomic Map