siRNA Details
| Gene Symbol: OTOR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020157.3 | NP_064542.1 | 4 | 605 | ||
| GenBank mRNA | AF233261.1 | - | 5 | 547 | ||
| AF243505.1 | - | 4 | 563 | |||
| AJ242552.1 | - | 4 | 556 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OTOR| Gene Symbol: OTOR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020157.3 | NP_064542.1 | 4 | 605 | ||
| GenBank mRNA | AF233261.1 | - | 5 | 547 | ||
| AF243505.1 | - | 4 | 563 | |||
| AJ242552.1 | - | 4 | 556 | |||