siRNA Details
| Gene Symbol: Slc39a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034929.1 | NP_001030101.1 | 5, 6 | 959 | ||
| XM_006240283.2 | XP_006240345.1 | 5, 6 | 1263 | |||
| XM_008764784.1 | XP_008763006.1 | 4, 5 | 1392 | |||
| GenBank mRNA | BC105849.1 | - | 5, 6 | 959 | 7457541 | |

Slc39a9
Genomic Map