siRNA Details
| Gene Symbol: Lrrc74 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037783.2 | NP_001032872.2 | 9 | 1321 | ||
| GenBank mRNA | BC083569.1 | - | 9 | 1270 | ||
| BC098927.1 | - | 1333 | ||||
| BC127444.1 | - | 9 | 1312 | 7135116 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrrc74| Gene Symbol: Lrrc74 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037783.2 | NP_001032872.2 | 9 | 1321 | ||
| GenBank mRNA | BC083569.1 | - | 9 | 1270 | ||
| BC098927.1 | - | 1333 | ||||
| BC127444.1 | - | 9 | 1312 | 7135116 | ||