siRNA Details
| Gene Symbol: Rnf126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033702.1 | NP_001028874.1 | 6, 7 | 694 | ||
| XM_006240899.1 | XP_006240961.1 | 6, 7 | 644 | |||
| GenBank mRNA | BC099810.1 | - | 6, 7 | 694 | 5621533 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnf126| Gene Symbol: Rnf126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033702.1 | NP_001028874.1 | 6, 7 | 694 | ||
| XM_006240899.1 | XP_006240961.1 | 6, 7 | 644 | |||
| GenBank mRNA | BC099810.1 | - | 6, 7 | 694 | 5621533 | |