siRNA Details
| Gene Symbol: C1qtnf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034932.1 | NP_001030104.1 | 3 | 1340 | ||
| GenBank mRNA | BC101907.1 | - | 3 | 1340 | 7933909 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

C1qtnf6| Gene Symbol: C1qtnf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034932.1 | NP_001030104.1 | 3 | 1340 | ||
| GenBank mRNA | BC101907.1 | - | 3 | 1340 | 7933909 | |