siRNA Details
Gene Symbol: Mettl7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037355.1 | NP_001032432.1 | 2 | 585 | ||
GenBank mRNA | BC104688.1 | - | 1 | 321 | ||
BC105760.1 | - | 2 | 585 | 5623503 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mettl7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037355.1 | NP_001032432.1 | 2 | 585 | ||
GenBank mRNA | BC104688.1 | - | 1 | 321 | ||
BC105760.1 | - | 2 | 585 | 5623503 |