siRNA Details
| Gene Symbol: Ncapd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034000.1 | NP_001029172.1 | 17 | 2134 | ||
| XM_006242735.2 | XP_006242797.1 | 17 | 2436 | |||
| XM_006242736.2 | XP_006242798.1 | 17 | 2087 | |||
| XM_008766053.1 | XP_008764275.1 | 16 | 2511 | |||
| GenBank mRNA | BC101928.1 | - | 17 | 2134 | 7939116 | |

Ncapd3
Genomic Map