siRNA Details
| Gene Symbol: Tmem45b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033067.1 | NP_001028239.1 | 6 | 1098 | ||
| GenBank mRNA | BC100635.1 | - | 6 | 1098 | 5621332 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem45b| Gene Symbol: Tmem45b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033067.1 | NP_001028239.1 | 6 | 1098 | ||
| GenBank mRNA | BC100635.1 | - | 6 | 1098 | 5621332 | |