siRNA Details
| Gene Symbol: Xylb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033704.1 | NP_001028876.1 | 19 | 2397 | ||
| XM_006244093.1 | XP_006244155.1 | 20 | 2773 | |||
| XM_006244094.1 | XP_006244156.1 | 19 | 2678 | |||
| XM_006244095.1 | XP_006244157.1 | 18 | 2590 | |||
| XM_006244096.1 | XP_006244158.1 | 18 | 2413 | |||
| GenBank mRNA | BC101852.1 | - | 19 | 2397 | 7930657 | |

Xylb
Genomic Map