siRNA Details
| Gene Symbol: Ddx21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037201.1 | NP_001032278.1 | 15 | 3644 | ||
| GenBank mRNA | BC105878.1 | - | 15 | 3644 | 7126328 | |
| Gene Symbol: LOC685025 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773110.1 | XP_008771332.1 | 2 | 3693 | ||

Ddx21
Genomic Map